9-Aminoacridine 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
9-AA
A196593-100G
A196593-1G
A196593-250MG
A196593-25G
A196593-5G
76721-314EA
504.99
USD
76721-314
76721-316
76721-318
76721-320
76721-322
9-Aminoacridine 98%
9-Aminoacridine
Precaución: for Research Use Only
Formula:
C₁₃H₁₀N₂ MW: 194.24 g/mol |
MDL Number:
MFCD00037839 CAS Number: 90-45-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...