4-Methylphenylboronic acid pinacol ester 98%

Supplier: Ambeed
Warning

Synonyms: 4,4,5,5-Tetramethyl-2-(p-tolyl)-1,3,2-dioxaborolane

A196359-100G A196359-10G A196359-1G A196359-250MG A196359-5G A196359-25G
76721-296EA 248 USD
76721-296 76721-298 76721-300 76721-302 76721-304 77600-598
4-Methylphenylboronic acid pinacol ester 98%
4-Methylphenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₁₉BO₂
MW: 218.1
MDL Number: MFCD03453662
CAS Number: 195062-57-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR