Methyl-5-nitrosalicylate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 2-hydroxy-5-nitrobenzoate

A196240-10G A196240-1G A196240-25G A196240-5G
76721-288EA 25.47 USD
76721-288 76721-290 76721-292 76721-294
Methyl-5-nitrosalicylate 98%
Methyl-5-nitrosalicylate

Precaución: for Research Use Only

Formula: C₈H₇NO₅
MW: 197.15 g/mol
MDL Number: MFCD00024695
CAS Number: 17302-46-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR