1-(3-Chlorobenzo[b]thiophen-2-yl)-N-methylmethanamine hydrochloride 98%

Supplier: Ambeed
Warning

A195755-1G A195755-250MG
76721-276EA 396.78 USD
76721-276 76721-278
1-(3-Chlorobenzo[b]thiophen-2-yl)-N-methylmethanamine hydrochloride 98%
1-(3-Chlorobenzo[b]thiophen-2-yl)-N-methylmethanamine hydrochloride

Precaución: for Research Use Only

Formula: C₁₀H₁₁Cl₂NS
MW: 248.17 g/mol
MDL Number: MFCD13193901
CAS Number: 132740-14-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR