Phthalimide DBU salt 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Phthalimide 1,8-Diazabicyclo[5.4.0]undec-7-ene Salt, DBU Phthalimide Salt
A195450-100G
A195450-25G
A195450-5G
76721-264EA
128
USD
76721-264
76721-266
76721-268
Phthalimide DBU salt 98%
Phthalimide DBU salt
Precaución: for Research Use Only
Formula:
C₁₇H₂₁N₃O₂ |
MDL Number:
MFCD00191683 CAS Number: 119812-51-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...