(Methyl)triphenylphosphonium bromide 98%
Supplier: Ambeed
|
Danger
|
A194648-100G
A194648-25G
A194648-500G
A194648-10G
76721-244EA
30
USD
76721-244
76721-246
76721-248
77567-826
(Methyl)triphenylphosphonium bromide 98%
(Methyl)triphenylphosphonium bromide
Precaución: For research use only.
Formula:
C₁₉H₁₈BrP MW: 357.23 g/mol Melting Pt: 232 |
MDL Number:
MFCD00011804 CAS Number: 1779-49-3 UN: 2811 ADR: 6.1 Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...