4-(4-Chlorophenyl)cyclohexanecarboxylic acid 98%

Supplier: Ambeed
Warning

A194353-100G A194353-10G A194353-500G A194353-5G A194353-25G
76721-230EA 62.6 USD
76721-230 76721-232 76721-234 76721-236 77667-628
4-(4-Chlorophenyl)cyclohexanecarboxylic acid 98%
4-(4-Chlorophenyl)cyclohexanecarboxylic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₅ClO₂
MW: 238.7100 g/mol
MDL Number: MFCD00665919
CAS Number: 95233-37-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR