2-Benzamido-2-phenylacetic acid 98%

Supplier: Ambeed
Warning

A193901-100G A193901-10G A193901-1G A193901-25G A193901-5G
76721-206EA 183.54 USD
76721-206 76721-208 76721-210 76721-212 76721-214
2-Benzamido-2-phenylacetic acid 98%
2-Benzamido-2-phenylacetic acid

Precaución: for Research Use Only

Formula: C₁₅H₁₃NO₃
MW: 255.26 g/mol
MDL Number: MFCD00666894
CAS Number: 29670-63-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR