Diethyl-1,4-dihydro-2,6-dimethyl-3,5-pyridinedicarboxylate 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Hantzsch Ester, 1,4-Dihydro-2,6-dimethyl-3,5-pyridinedicarboxylic Acid Diethyl Ester, Diethyl 1,4-dihydro-3,5-pyridinecarboxylate, Etidin, Diethyl 1,4-dihydro-2,6-dimethyl-3,5-pyridinedicarboxylate
A193361-100G
A193361-25G
A193361-500G
76721-186EA
9.55
USD
76721-186
76721-188
76721-190
Diethyl-1,4-dihydro-2,6-dimethyl-3,5-pyridinedicarboxylate 97%
Diethyl-1,4-dihydro-2,6-dimethyl-3,5-pyridinedicarboxylate
Precaución: for Research Use Only
Formula:
C₁₃H₁₉NO₄ MW: 253.30 g/mol |
MDL Number:
MFCD00005951 CAS Number: 1149-23-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...