Rivastigmine-L-tartrate 98%

Supplier: Ambeed
Warning

Synonyms: Rivastigmine tartrate, (S)-3-(1-(Dimethylamino)ethyl)phenyl ethyl(methyl)carbamate (2R,3R)-2,3-dihydroxysuccinate, N-Ethyl-N-methylcarbamic Acid 3-[(S)-1-(Dimethylamino)ethyl]phenyl Ester L-Tartrate, 3-[(S)-1-(Dimethylamino)ethyl]phenyl N-Ethyl-N-methylcarbamate L-Tartrate

A193069-100MG A193069-10MG A193069-1G A193069-25G A193069-50MG A193069-5G A193069-250MG
76721-174EA 10 USD
76721-174 76721-176 76721-178 76721-180 76721-182 76721-184 77600-566
Rivastigmine-L-tartrate 98%
Rivastigmine-L-tartrate

Precaución: for Research Use Only

Formula: C₁₄H₂₂N₂O₂·C₄H₆O₆
MW: 400.43 g/mol
MDL Number: MFCD03700731
CAS Number: 129101-54-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR