Methyl-4-methoxy-3-nitrobenzoate 98%

Supplier: Ambeed
Warning

Synonyms: 4-Methoxy-3-nitrobenzoic acid methyl ester, Methyl 4-methoxy-3-nitrobenzoate

A192888-1G A192888-25G A192888-5G
76721-164EA 15 USD
76721-164 76721-166 76721-168
Methyl-4-methoxy-3-nitrobenzoate 98%
Methyl-4-methoxy-3-nitrobenzoate

Precaución: for Research Use Only

Formula: C₉H₉NO₅
MDL Number: MFCD00024213
CAS Number: 40757-20-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR