Boc-5-aminolevulinic acid 96%

Supplier: Ambeed
Warning

Synonyms: 5-((tert-Butoxycarbonyl)amino)-4-oxopentanoic acid

A191519-250MG A191519-1G
76721-136EA 100.99 USD
76721-136 77550-228
Boc-5-aminolevulinic acid 96%
Boc-5-aminolevulinic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₇NO₅
MDL Number: MFCD02682635
CAS Number: 72072-06-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR