N,N-Diisopropylethylenediamine 97%

Supplier: Ambeed
Danger

Synonyms: 2-Aminoethyldiisopropylamine, N,N-Bis(isopropyl)ethane-1,2-diamine

A191357-25G A191357-5G
76721-132EA 18.04 USD
76721-132 76721-134
N,N-Diisopropylethylenediamine 97%
N,N-Diisopropylethylenediamine

Precaución: for Research Use Only

Formula: C₈H₂₀N₂
MW: 144.26 g/mol
MDL Number: MFCD00015015
CAS Number: 121-05-1
UN: 2734
ADR: 8,3,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR