N-(4-Bromophenyl)phthalimide 98%

Supplier: Ambeed
Warning

Synonyms: 2-(4-Bromophenyl)isoindoline-1,3-dione

A190332-5G A190332-1G
76721-118EA 51.01 USD
76721-118 77879-217
N-(4-Bromophenyl)phthalimide 98%
N-(4-Bromophenyl)phthalimide

Precaución: for Research Use Only

Formula: C₁₄H₈BrNO₂
MDL Number: MFCD00047301
CAS Number: 40101-31-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR