Monoethyl succinate 98%

Supplier: Ambeed
Warning

Synonyms: Ethyl hydrogen succinate, 4-Ethoxy-4-oxobutanoic acid, mono-Ethyl succinate

A190138-25G A190138-5G A190138-10G A190138-100G
76721-114EA 24 USD
76721-114 76721-116 77600-542 77879-216
Monoethyl succinate 98%
Monoethyl succinate

Precaución: for Research Use Only

Formula: C₆H₁₀O₄
MW: 146.14 g/mol
MDL Number: MFCD00053346
CAS Number: 1070-34-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR