Ethyl-5-amino-1-phenyl-1H-pyrazole-4-carboxylate 98%

Supplier: Ambeed
Warning

Synonyms: 5-Amino-1-phenylpyrazole-4-carboxylic acid Ethyl Ester, 5-Amino-1-phenyl-1H-pyrazole-4-carboxylic acid ethyl ester, 5-Amino-1-phenylpyrazole-4-carboxylic acid ethyl ester, 5-Amino-4-carbethoxy-1-phenylpyrazole, Ethyl 5-Amino-1-phenylpyrazole-4-carboxylate, Ethyl 5-amino-1-phenyl-1H-pyrazole-4-carboxylate

A189228-10G A189228-1G A189228-250MG A189228-25G A189228-5G
76721-096EA 43.5 USD
76721-096 76721-098 76721-100 76721-102 76721-104
Ethyl-5-amino-1-phenyl-1H-pyrazole-4-carboxylate 98%
Ethyl 5-amino-1-phenyl-1H-pyrazole-4-carboxylate

Precaución: for Research Use Only

Formula: C₇H₁₁N₃O₂
MW: 231.26 g/mol
MDL Number: MFCD00020731
CAS Number: 16078-71-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR