N-(4-(Aminomethyl)phenyl)acetamide hydrochloride 95%

Supplier: Ambeed
Warning

A188399-100MG A188399-250MG A188399-1G
76721-076EA 29.71 USD
76721-076 76721-078 77667-568
N-(4-(Aminomethyl)phenyl)acetamide hydrochloride 95%
N-(4-(Aminomethyl)phenyl)acetamide hydrochloride

Precaución: for Research Use Only

Formula: C₉H₁₃ClN₂O
MW: 200.67 g/mol
MDL Number: MFCD06656043
CAS Number: 25027-73-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR