m-Tolidine 97%

Supplier: Ambeed
Warning

Synonyms: 2,2'-Dimethyl-[1,1'-biphenyl]-4,4'-diamine, 4,4'-Diamino-2,2'-dimethylbiphenyl, 2,2'-Dimethylbenzidine

A187639-100G A187639-25G A187639-5G A187639-500G A187639-10G
76721-058EA 65.78 USD
76721-058 76721-060 76721-062 77567-748 77667-562
m-Tolidine 97%
m-Tolidine

Precaución: For research use only.

Formula: C₁₄H₁₆N₂
MW: 212.30 g/mol
MDL Number: MFCD02269175
CAS Number: 84-67-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR