Potassium-D-gluconate 99%

Supplier: Ambeed
Warning

Synonyms: Potassium Gluconate, Gluconic Acid Potassium Salt, D-Gluconic acid potassium salt, Potassium (2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate

A186454-100G A186454-25G A186454-500G
76721-026EA 7.43 USD
76721-026 76721-028 77600-510
Potassium-D-gluconate 99%
Potassium-D-gluconate

Precaución: for Research Use Only

Formula: HOH₂C(CH(OH))₄COOK
MW: 234.25 g/mol
MDL Number: MFCD00064211
CAS Number: 299-27-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR