Methyl-2-ethoxybenzoate 95+%
Supplier: Ambeed
|
Warning
|
Synonyms:
Methyl 2-ethoxybenzoate
A186010-1G
A186010-25G
A186010-5G
A186010-100G
76721-020EA
9
USD
76721-020
76721-022
76721-024
77879-200
Methyl-2-ethoxybenzoate 95+%
Methyl 2-ethoxybenzoate
Precaución: for Research Use Only
Formula:
C₁₀H₁₂O₃ |
MDL Number:
MFCD00017261 CAS Number: 3686-55-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...