Methyl-4-hydroxy-3-(trifluoromethyl)benzoate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 4-hydroxy-3-(trifluoromethyl)benzoate

A185772-1G A185772-25G A185772-5G A185772-10G
76721-018EA 14 USD
76721-018 77667-548 77879-199 77879-198
Methyl-4-hydroxy-3-(trifluoromethyl)benzoate 98%
Methyl-4-hydroxy-3-(trifluoromethyl)benzoate

Precaución: for Research Use Only

Formula: C₉H₇F₃O₃
MDL Number: MFCD06204091
CAS Number: 115933-50-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR