Ethyl (2-Methylbenzoyl)acetate 95%

Supplier: Ambeed
Warning

A184835-1G A184835-25G A184835-5G
76721-000EA 22 USD
76721-000 76721-002 76721-004
Ethyl (2-Methylbenzoyl)acetate 95%
Ethyl (2-Methylbenzoyl)acetate

Precaución: for Research Use Only

Formula: C₁₂H₁₄O₃
MDL Number: MFCD03424817
CAS Number: 51725-82-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR