Methyl isobutyrylacetate 95%

Supplier: Ambeed
Warning

Synonyms: Methyl 4-methyl-3-oxopentanoate, Methyl 4-methyl-3-oxovalerate

A182738-10G A182738-500G A182738-100G A182738-1000G
76720-956EA 5 USD
76720-956 76720-958 77567-678 77667-520
Methyl isobutyrylacetate 95%
Methyl isobutyrylacetate

Precaución: For research use only.

Formula: C₇H₁₂O₃
MW: 144.17 g/mol
MDL Number: MFCD00040499
CAS Number: 42558-54-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR