2,3,6-Trichloroaniline 95%

Supplier: Ambeed
Warning

A182287-100MG A182287-1G A182287-250MG A182287-5G
76720-930EA 23.34 USD
76720-930 76720-932 76720-934 76720-936
2,3,6-Trichloroaniline 95%
2,3,6-Trichloroaniline

Precaución: for Research Use Only

Formula: C₆H₄Cl₃N
MW: 196.46 g/mol
MDL Number: MFCD02683886
CAS Number: 88963-39-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR