4-Benzyloxy-2-fluorophenylboronic acid 97%

Supplier: Ambeed
Warning

Synonyms: 4-Benzyloxy-2-fluorobenzeneboronic acid, (4-(Benzyloxy)-2-fluorophenyl)boronic acid

A181077-10G A181077-1G A181077-250MG A181077-5G
76720-876EA 210.06 USD
76720-876 76720-878 76720-880 76720-882
4-Benzyloxy-2-fluorophenylboronic acid 97%
4-Benzyloxy-2-fluorophenylboronic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₂BFO₃
MW: 246.04 g/mol
MDL Number: MFCD02093064
CAS Number: 166744-78-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR