4-Biphenylylmethanol 95%

Supplier: Ambeed
Warning

Synonyms: Biphenyl-4-methanol, [1,1'-Biphenyl]-4-ylmethanol, 4-Phenylbenzyl alcohol, 4-Biphenylmethanol

A180856-100G A180856-10G A180856-25G A180856-500G
76720-868EA 48 USD
76720-868 76720-870 76720-872 76720-874
4-Biphenylylmethanol 95%
4-Biphenylylmethanol

Precaución: for Research Use Only

Formula: C₁₃H₁₂O
MW: 184.24 g/mol
MDL Number: MFCD00004660
CAS Number: 3597-91-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR