Cyclohexyl-p-toluenesulfonate 97%
Supplier: Ambeed
|
Warning
|
A180718-1G
A180718-25G
A180718-5G
76720-862EA
11
USD
76720-862
76720-864
76720-866
Cyclohexyl-p-toluenesulfonate 97%
Cyclohexyl-p-toluenesulfonate
Precaución: for Research Use Only
Formula:
C₁₃H₁₈O₃S |
MDL Number:
MFCD00014291 CAS Number: 953-91-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...