1,3,2-Dioxathiane-2-oxide 98%
Supplier: Ambeed
|
Danger
|
Synonyms:
1,3,2-Dioxathiane 2-oxide, 1,3-Propanediol Cyclic sulphite, Trimethylene sulphite, 1,3-Propylene Sulphite
A180001-10G
A180001-25G
A180001-5G
76720-848EA
9.55
USD
76720-848
76720-850
76720-852
1,3,2-Dioxathiane-2-oxide 98%
1,3,2-Dioxathiane-2-oxide
Precaución: for Research Use Only
Formula:
C₃H₆O₃S MW: 122.14 g/mol |
MDL Number:
MFCD01682880 CAS Number: 4176-55-0 UN: 1760 ADR: 8,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...