Nonadecanedioic acid 96%

Supplier: Ambeed
Warning

A179904-1G A179904-250MG A179904-5G
76720-842EA 26 USD
76720-842 76720-844 76720-846
Nonadecanedioic acid 96%
Nonadecanedioic acid

Precaución: for Research Use Only

Formula: C₁₉H₃₆O₄
MDL Number: MFCD00039536
CAS Number: 6250-70-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR