5-Methoxy-2-methyl-3-indoleacetic acid 98%

Supplier: Ambeed
Warning

Synonyms: Indomethacin Related Compound A

A179655-100MG A179655-1G A179655-250MG A179655-5G A179655-25G
76720-826EA 12.74 USD
76720-826 76720-828 76720-830 76720-832 77667-484
5-Methoxy-2-methyl-3-indoleacetic acid 98%
5-Methoxy-2-methyl-3-indoleacetic acid

Precaución: for Research Use Only

Formula: C₁₂H₁₃NO₃
MW: 219.24 g/mol
MDL Number: MFCD00005618
CAS Number: 2882-15-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR