(4-Chlorophenyl)acetic acid methyl ester 98%

Supplier: Ambeed
Warning

Synonyms: (4-Chlorophenyl)acetic acid methyll ester, Methyl (4-chlorophenyl)acetate, Methyl (4-Chlorophenyl)acetate, Methyl 4-chlorophenylacetate

A179349-100G A179349-10G A179349-25G A179349-500G A179349-5G
76720-812EA 63.66 USD
76720-812 76720-814 76720-816 76720-818 76720-820
(4-Chlorophenyl)acetic acid methyl ester 98%
(4-Chlorophenyl)acetic acid methyl ester

Precaución: for Research Use Only

Formula: (C₆H₄Cl)CH₂COOCH₃
MW: 184.62 g/mol
MDL Number: MFCD00032743
CAS Number: 52449-43-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR