Methyl-2-(1,3-dioxo-1,3-dihydro-2H-isoindol-2-yl)acetate 98%
Supplier: Ambeed
|
Danger
|
Synonyms:
Methyl phthalimid-1-ylacetate
A179079-10G
A179079-1G
A179079-25G
A179079-5G
76720-802EA
135
USD
76720-802
76720-804
76720-806
76720-808
Methyl-2-(1,3-dioxo-1,3-dihydro-2H-isoindol-2-yl)acetate 98%
Methyl-2-(1,3-dioxo-1,3-dihydro-2H-isoindol-2-yl)acetate
Precaución: for Research Use Only
Formula:
C₁₁H₉NO₄ |
MDL Number:
MFCD00158668 CAS Number: 23244-58-8 UN: 2811 ADR: 6.1,Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...