2-(1-tert-Butoxycarbonylazetidin-3-yl)acetic acid 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-[1-Boc-azetidin-3-yl]acetic acid, 2-[1-(tert-Butoxycarbonyl)azetidin-3-yl]acetic acid
A178845-100G
A178845-10G
A178845-1G
A178845-250MG
A178845-25G
A178845-5G
76720-790EA
440.28
USD
76720-790
76720-792
76720-794
76720-796
76720-798
76720-800
2-(1-tert-Butoxycarbonylazetidin-3-yl)acetic acid 97%
2-(1-tert-Butoxycarbonylazetidin-3-yl)acetic acid
Precaución: for Research Use Only
Formula:
C₁₀H₁₇NO₄ MW: 215.25 g/mol |
MDL Number:
MFCD02179028 CAS Number: 183062-96-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...