N-(Carbobenzoxy)-1H-pyrazole-1-carboxamidine 98%

Supplier: Ambeed
Warning

Synonyms: N-(Benzyloxycarbonyl)-1H-pyrazole-1-carboxamidine, 1-(N-Cbz-amidino)pyrazole, 1-[N-(Carbobenzoxy)amidino]pyrazole, N-Cbz-1H-pyrazole-1-carboxamidine, Benzyl (imino(1H-pyrazol-1-yl)methyl)carbamate

A178404-100G A178404-10G A178404-1G A178404-25G A178404-5G
76720-774EA 342 USD
76720-774 76720-776 76720-778 76720-780 76720-782
N-(Carbobenzoxy)-1H-pyrazole-1-carboxamidine 98%
N-(Carbobenzoxy)-1H-pyrazole-1-carboxamidine

Precaución: for Research Use Only

Formula: C₁₂H₁₂N₄O₂
MW: 244.25 g/mol
MDL Number: MFCD00274657
CAS Number: 152120-62-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR