Probucol 98%

Supplier: Ambeed
Warning

A177844-100G A177844-100MG A177844-1G A177844-25G A177844-25MG A177844-5G
76720-752EA 165 USD
76720-752 76720-754 76720-756 76720-758 76720-760 76720-762
Probucol 98%
Probucol

Precaución: for Research Use Only

Formula: C₃₁H₄₈O₂S₂
MDL Number: MFCD00079281
CAS Number: 23288-49-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR