2-Methoxy-3-pyridineboronic acid pinacol ester 98%

Supplier: Ambeed
Warning

Synonyms: 2-Methoxypyridine-3-boronic acid pinacol ester, 2-Methoxy-3-(4,4,5,5-tetramethyl-[1,3,2]dioxaborolan-2-yl)-pyridine, 2-Methoxy-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

A177722-1G A177722-25G A177722-5G
76720-746EA 11 USD
76720-746 76720-748 76720-750
2-Methoxy-3-pyridineboronic acid pinacol ester 98%
2-Methoxy-3-pyridineboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₂H₁₈BNO₃
MDL Number: MFCD05663507
CAS Number: 532391-31-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR