Propyl dodecanoate 97%
Supplier: Ambeed
|
Warning
|
A177718-10G
A177718-1G
A177718-25G
A177718-5G
A177718-100G
76720-738EA
23
USD
76720-738
76720-740
76720-742
76720-744
77879-172
Propyl dodecanoate 97%
Propyl dodecanoate
Precaución: for Research Use Only
Formula:
C₁₅H₃₀O₂ |
MDL Number:
MFCD00056190 CAS Number: 3681-78-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...