Methyl 3-(3-hydroxyphenyl)propionate 95%

Supplier: Ambeed
Warning

A177481-250MG A177481-1G
76720-726EA 55.17 USD
76720-726 77667-466
Methyl 3-(3-hydroxyphenyl)propionate 95%
Methyl 3-(3-hydroxyphenyl)propionate

Precaución: for Research Use Only

Formula: C₁₀H₁₂O₃
MW: 180.2 g/mol
MDL Number: MFCD03788460
CAS Number: 61389-68-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR