Potassium(morpholin-4-ylmethyl)trifluoroborate 95%

Supplier: Ambeed
Warning

Synonyms: Potassium trifluoro(morpholinomethyl)borate

A177377-1G A177377-250MG
76720-722EA 23 USD
76720-722 76720-724
Potassium(morpholin-4-ylmethyl)trifluoroborate 95%
Potassium (morpholin-4-ylmethyl)trifluoroborate

Precaución: for Research Use Only

Formula: C₅H₁₀BF₃KNO
MW: 207.0435 g/mol
MDL Number: MFCD10566512
CAS Number: 936329-94-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR