N-tert-Butylglycine hydrochloride 97%

Supplier: Ambeed
Warning

A177263-100G A177263-10G A177263-25G A177263-500G A177263-5G
76720-712EA 71.01 USD
76720-712 76720-714 76720-716 76720-718 76720-720
N-tert-Butylglycine hydrochloride 97%
N-tert-Butylglycine hydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₄ClNO₂
MDL Number: MFCD03788441
CAS Number: 6939-23-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR