Methyl-2-methyl-3-furoate 95%

Supplier: Ambeed
Warning

Synonyms: Methyl 2-methyl-3-furoate

A176800-1G A176800-5G A176800-100G A176800-25G
76720-704EA 12 USD
76720-704 76720-706 77879-165 77879-166
Methyl-2-methyl-3-furoate 95%
Methyl-2-methyl-3-furoate

Precaución: for Research Use Only

Formula: C₇H₈O₃
MW: 140.14
MDL Number: MFCD00003249
CAS Number: 6141-58-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR