Phenazine 97%

Supplier: Ambeed
Warning

A176655-100G A176655-10G A176655-1G A176655-25G A176655-5G
76720-694EA 89.12 USD
76720-694 76720-696 76720-698 76720-700 76720-702
Phenazine 97%
Phenazine

Precaución: for Research Use Only

Formula: C₁₂H₈N₂
MW: 180.21 g/mol
MDL Number: MFCD00005023
CAS Number: 92-82-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR