(R)-2-Benzamido-2-phenylacetic acid 97%

Supplier: Ambeed
Warning

Synonyms: N-D-2-Bz-Phg-OH

A176204-1G A176204-250MG
76720-684EA 39.26 USD
76720-684 76720-686
(R)-2-Benzamido-2-phenylacetic acid 97%
(R)-2-Benzamido-2-phenylacetic acid

Precaución: for Research Use Only

Formula: C₁₅H₁₃NO₃
MW: 255.26 g/mol
MDL Number: MFCD02684297
CAS Number: 10419-67-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR