Piperazine-1-carboxylic acid amide hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: Piperazine-1-carboxamide hydrochloride

A176197-100MG A176197-1G A176197-250MG
76720-678EA 23.34 USD
76720-678 76720-680 76720-682
Piperazine-1-carboxylic acid amide hydrochloride 97%
Piperazine-1-carboxylic acid amide hydrochloride

Precaución: for Research Use Only

Formula: C₅H₁₂ClN₃O
MW: 165.62 g/mol
MDL Number: MFCD06408948
CAS Number: 474711-89-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR