Sodium tetrachloropalladate(II) 98%

Supplier: Ambeed
Warning

Synonyms: Palladium sodium chloride, Sodium palladium (II) chloride, Disodium tetrachloropalladate

A176160-10G A176160-1G A176160-250MG A176160-25G A176160-5G
76720-668EA 674.74 USD
76720-668 76720-670 76720-672 76720-674 76720-676
Sodium tetrachloropalladate(II) 98%
Sodium tetrachloropalladate(II)

Precaución: for Research Use Only

Formula: Cl₄Na₂Pd
MW: 294.21 g/mol
MDL Number: MFCD00003487
CAS Number: 13820-53-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR