N-Ethylmaleimide (NEM) 97%

Supplier: Ambeed
Danger

Synonyms: N-Ethylmaleimide, 1-Ethyl-1H-pyrrole-2,5-dione

A175883-100G A175883-1G A175883-25G A175883-5G
76720-660EA 392 USD
76720-660 76720-662 76720-664 76720-666
N-Ethylmaleimide (NEM) 97%
N-Ethylmaleimide (NEM)

Precaución: for Research Use Only

Formula: C₆H₇NO₂
MDL Number: MFCD00005509
CAS Number: 128-53-0
UN: 2928
ADR: 6.1,8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR