3,4-(Methylenedioxy)phenylboronic acid pinacol ester 95%

Supplier: Ambeed
Warning

Synonyms: 5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-benzo-1,3-dioxole, 2-(Benzo[d][1,3]dioxol-5-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 3,4-Ethylenedioxybenzene boronic acid, pinacol ester

A175613-10G A175613-1G A175613-250MG A175613-25G A175613-5G
76720-644EA 113.52 USD
76720-644 76720-646 76720-648 76720-650 77593-112
3,4-(Methylenedioxy)phenylboronic acid pinacol ester 95%
3,4-(Methylenedioxy)phenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₁₇BO₄
MW: 248.0827 g/mol
MDL Number: MFCD05663889
CAS Number: 94838-82-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR