Nepafenac 98%

Supplier: Ambeed
Danger

Synonyms: 2-Amino-3-benzoylbenzeneacetamide, 2-(2-Amino-3-benzoylphenyl)acetamide

A174101-100MG A174101-10MG A174101-1G A174101-1MG A174101-250MG A174101-25MG A174101-50MG A174101-5G
76720-610EA 61.54 USD
76720-610 76720-612 76720-614 76720-616 76720-618 76720-620 76720-622 77667-424
Nepafenac 98%
Nepafenac

Precaución: for Research Use Only

Formula: C₁₅H₁₄N₂O₂
MW: 254.29 g/mol
MDL Number: MFCD08067732
CAS Number: 78281-72-8
UN: 3077
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR