Sodium trifluoroacetate 98%

Supplier: Ambeed
Warning

Synonyms: Sodium 2,2,2-trifluoroacetate

A173394-1000G A173394-100G A173394-10G A173394-25G A173394-500G
76720-584EA 110.34 USD
76720-584 76720-586 76720-588 76720-590 76720-592
Sodium trifluoroacetate 98%
Sodium trifluoroacetate

Precaución: for Research Use Only

Formula: C₂F₃NaO₂
MW: 136.01 g/mol
MDL Number: MFCD00013217
CAS Number: 2923-18-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR