4-Nitrophenyl butyrate 95%

Supplier: Ambeed
Warning

A172713-250MG A172713-5G A172713-1G A172713-25G
76720-562EA 11.67 USD
76720-562 76720-564 77600-394 77600-396
4-Nitrophenyl butyrate 95%
4-Nitrophenyl butyrate

Precaución: for Research Use Only

Formula: C₁₀H₁₁NO₄
MW: 209.19 g/mol
MDL Number: MFCD00047734
CAS Number: 2635-84-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR